Assay Details
Target Gene Details
Entrez Gene ID: | 4683 |
Gene Name: | nibrin |
Gene Aliases: |
AT-V1, AT-V2, ATV, NBS, NBS1, P95 |
Location: |
Chr.8:89933336-89984733 on Build GRCh38 |
Assay Gene Location: | Within Intron 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NBN | NM_001024688.2 | NP_001019859.1 | ||
| NM_002485.4 | NP_002476.2 | |||
| XM_011517045.1 | XP_011515347.1 | |||
| XM_017013460.1 | XP_016868949.1 | |||
| XM_017013461.1 | XP_016868950.1 | |||
| XM_017013462.1 | XP_016868951.1 | |||
| AF051334.1 | AAC39732.1 | |||
| AF058696.2 | AAC39752.1 | |||
| AK223256.1 | BAD96976.1 | |||
| AK289848.1 | ||||
| AK312410.1 | ||||
| BC136802.1 | ||||
| BC136803.1 | ||||
| BC146797.1 | ||||
| BX640816.1 | CAH56160.1 |
More Information
Set Membership: |
|

Genomic Map