Assay Details
Target Gene Details
Entrez Gene ID: | 7976 |
Gene Name: | frizzled class receptor 3 |
Gene Aliases: |
Fz-3 |
Location: |
Chr.8:28494205-28574268 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FZD3 | NM_017412.3 | NP_059108.1 | ||
| NM_145866.1 | NP_665873.1 | |||
| XM_017013842.1 | XP_016869331.1 | |||
| XM_017013843.1 | XP_016869332.1 | |||
| XM_017013844.1 | XP_016869333.1 | |||
| AB039723.1 | BAA94968.1 | |||
| AJ272427.1 | CAB89114.1 | |||
| AK291480.1 | ||||
| AY005130.3 | AAF89088.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv7157n100 | Chr.8:28499087 - 28584581 on Build GRCh38 | Loss |
|
| nsv1015224 | Chr.8:28504799 - 28549099 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs79373076] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map