Assay Details
Target Gene Details
Entrez Gene ID: | 1808 |
Gene Name: | dihydropyrimidinase like 2 |
Gene Aliases: |
CRMP-2, CRMP2, DHPRP2, DRP-2, DRP2, N2A3, ULIP-2, ULIP2 |
Location: |
Chr.8:26514193-26658177 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DPYSL2 | NM_001197293.2 | NP_001184222.1 | ||
| AB209195.1 | BAD92432.1 | |||
| AB370195.1 | ||||
| AK092377.1 | BAC03872.1 | |||
| DA649686.1 |
More Information
Set Membership: |
|

Genomic Map