Assay Details
Target Gene Details
Entrez Gene ID: | 5862 |
Gene Name: | RAB2A, member RAS oncogene family |
Gene Aliases: |
LHX, RAB2 |
Location: |
Chr.8:60516910-60623644 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAB2A | NM_001242644.1 | 1 | 122 | NP_001229573.1 |
| NM_002865.2 | 1 | 122 | NP_002856.1 | |
| AK297576.1 | 1 | 96 | ||
| AK312344.1 | ||||
| BC008929.2 | AAH08929.1 | |||
| DA648682.1 | 1 | 124 | ||
| X12953.1 | CAA31411.1 |
More Information
Set Membership: |
|

Genomic Map