Assay Details
Target Gene Details
Entrez Gene ID: | 100616115 |
Gene Name: | microRNA 4469 |
Gene Aliases: |
mir-4469 |
Location: |
Chr.8:42896197-42896275 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MIR4469 | NR_039679.1 |
Target Gene Details
Entrez Gene ID: | 81790 |
Gene Name: | ring finger protein 170 |
Gene Aliases: |
ADSA, SNAX1 |
Location: |
Chr.8:42849637-42897288 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RNF170 | NM_001160223.1 | NP_001153695.1 | ||
| NM_001160224.1 | NP_001153696.1 | |||
| NM_001160225.1 | NP_001153697.1 | |||
| NM_030954.3 | NP_112216.3 | |||
| NR_027668.1 | ||||
| NR_027669.1 | ||||
| XM_006716404.2 | XP_006716467.1 | |||
| XM_006716405.2 | XP_006716468.1 | |||
| XM_011544666.2 | XP_011542968.1 | |||
| XM_017013881.1 | XP_016869370.1 | |||
| XM_017013882.1 | XP_016869371.1 | |||
| AK027748.1 | BAB55340.1 | |||
| AK090864.1 | BAC03534.1 | |||
| AK225086.1 | ||||
| AK225568.1 | ||||
| AL136620.1 | CAB66555.1 | |||
| BC013422.1 | AAH13422.1 | |||
| BC032393.1 | AAH32393.1 | |||
| BC039461.1 | AAH39461.1 | |||
| BC044566.1 | AAH44566.1 | |||
| BG910500.1 | ||||
| DA751961.1 |
More Information
Set Membership: |
|

Genomic Map