Assay Details
Target Gene Details
Entrez Gene ID: | 51115 |
Gene Name: | regulator of microtubule dynamics 1 |
Gene Aliases: |
CGI-90, FAM82B, RMD-1, RMD1 |
Location: |
Chr.8:86467116-86514348 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RMDN1 | NM_001317807.1 | NP_001304736.1 | ||
| XM_005250948.3 | XP_005251005.1 | |||
| XM_017013524.1 | XP_016869013.1 | |||
| XM_017013525.1 | XP_016869014.1 | |||
| XM_017013526.1 | XP_016869015.1 | |||
| XM_017013528.1 | XP_016869017.1 | |||
| AL561197.3 | ||||
| BU932433.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1023510 | Chr.8:86416772 - 86862720 on Build GRCh38 | Loss |
|

Genomic Map