Assay Details
Target Gene Details
Entrez Gene ID: | 101410533 |
Gene Name: | YTHDF3 antisense RNA 1 (head to head) |
Gene Aliases: |
- |
Location: |
Chr.8:63167725-63168442 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| YTHDF3-AS1 | NR_102684.1 | |||
| BM967225.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv611448 | Chr.8:63038678 - 63167927 on Build GRCh38 | Loss |
|

Genomic Map