Assay Details
Target Gene Details
Entrez Gene ID: | 83877 |
Gene Name: | TM2 domain containing 2 |
Gene Aliases: |
BLP1 |
Location: |
Chr.8:38988808-38997139 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TM2D2 | NM_001024380.1 | NP_001019551.1 | ||
| NM_001024381.1 | NP_001019552.1 | |||
| NM_031940.3 | NP_114146.3 | |||
| NM_078473.2 | NP_510882.1 | |||
| XM_005273657.3 | XP_005273714.1 | |||
| XM_006716408.3 | XP_006716471.1 | |||
| XM_011544667.2 | XP_011542969.1 | |||
| AF353991.1 | AAK35065.1 | |||
| AK075027.1 | BAC11359.1 | |||
| AK075155.1 | BAC11437.1 | |||
| AK314703.1 | ||||
| AL834224.1 | CAH10695.1 | |||
| BC004878.2 | ||||
| BC109049.1 | AAI09050.1 | |||
| BC109050.1 | AAI09051.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv820251 | Chr.8:38991221 - 38995507 on Build GRCh38 | Loss |
|

Genomic Map