Assay Details
Target Gene Details
Entrez Gene ID: | 100507056 |
Gene Name: | colon cancer associated transcript 1 (non-protein coding) |
Gene Aliases: |
CARLo-5, onco-lncRNA-40 |
Location: |
Chr.8:127207382-127219268 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCAT1 | NR_108049.1 | 2 | 2552 | |
| HM358356.1 | 2 | 2150 | ||
| HY370674.1 | 1 | 244 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831457 | Chr.8:127083126 - 127260605 on Build GRCh38 | Gain |
|

Genomic Map