Assay Details
Target Gene Details
Entrez Gene ID: | 55529 |
Gene Name: | transmembrane protein 55A |
Gene Aliases: |
- |
Location: |
Chr.8:90994271-91040975 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM55A | NM_018710.2 | NP_061180.1 | ||
| XM_011517152.2 | XP_011515454.1 | |||
| AK313783.1 | ||||
| AX775961.1 | CAE11680.1 | |||
| BC033892.1 | AAH33892.1 | |||
| CR749733.1 | CAH18492.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759629 | Chr.8:90962271 - 91261496 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map