Assay Details
Target Gene Details
Entrez Gene ID: | 11075 |
Gene Name: | stathmin 2 |
Gene Aliases: |
SCG10, SCGN10 |
Location: |
Chr.8:79610814-79666175 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STMN2 | NM_001199214.1 | NP_001186143.1 | ||
| NM_007029.3 | NP_008960.2 | |||
| AK292737.1 | ||||
| AK297485.1 | ||||
| AK297594.1 | ||||
| AK299500.1 | ||||
| AK308557.1 | ||||
| BC006302.2 | AAH06302.1 | |||
| BQ069488.1 | ||||
| D50375.1 | BAA23326.1 | |||
| DC319376.1 | ||||
| S82024.1 | AAB36428.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1075149 | Chr.8:79611264 - 79612465 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map