Assay Details
Target Gene Details
Entrez Gene ID: | 3612 |
Gene Name: | inositol monophosphatase 1 |
Gene Aliases: |
IMP, IMPA |
Location: |
Chr.8:81656916-81686354 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IMPA1 | NM_001144878.1 | NP_001138350.1 | ||
| NM_001144879.1 | NP_001138351.1 | |||
| NM_005536.3 | NP_005527.1 | |||
| AF042729.2 | AAB97468.1 | |||
| AK297078.1 | ||||
| AK300750.1 | ||||
| AK312823.1 | ||||
| BC008381.1 | AAH08381.1 | |||
| BC009565.1 | AAH09565.1 | |||
| X66922.1 | CAA47359.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv611607 | Chr.8:81405621 - 81768032 on Build GRCh38 | Gain |
|
| nsv831369 | Chr.8:81605833 - 81823586 on Build GRCh38 | Gain+Loss |
|
| nsv831368 | Chr.8:81554926 - 81697395 on Build GRCh38 | Gain |
|
| esv2737230 | Chr.8:81633446 - 81689714 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map