Assay Details
Target Gene Details
Entrez Gene ID: | 157769 |
Gene Name: | family with sequence similarity 91 member A1 |
Gene Aliases: |
- |
Location: |
Chr.8:123768439-123815452 on Build GRCh38 |
Assay Gene Location: | Within Intron 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM91A1 | NM_001317917.1 | NP_001304846.1 | ||
| NM_001317918.1 | NP_001304847.1 | |||
| NM_144963.3 | NP_659400.2 | |||
| AK074370.1 | BAB85063.1 | |||
| AK300603.1 | ||||
| AL832830.1 | CAH56231.1 | |||
| BC030520.1 | AAH30520.1 | |||
| DQ228141.1 | ABB17070.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831451 | Chr.8:123698706 - 123872908 on Build GRCh38 | Gain+Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs72715039] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map