Assay Details
Target Gene Details
Entrez Gene ID: | 51571 |
Gene Name: | family with sequence similarity 49 member B |
Gene Aliases: |
BM-009, L1 |
Location: |
Chr.8:129839593-130016697 on Build GRCh38 |
Assay Gene Location: | Within Intron 18 |
More Information
Set Membership: |
|

Genomic Map