Assay Details
Target Gene Details
Entrez Gene ID: | 84955 |
Gene Name: | NudC domain containing 1 |
Gene Aliases: |
CML66, OVA66 |
Location: |
Chr.8:109240919-109334205 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NUDCD1 | NM_001128211.1 | NP_001121683.1 | ||
| NM_032869.3 | NP_116258.2 | |||
| XM_017013910.1 | XP_016869399.1 | |||
| XM_017013911.1 | XP_016869400.1 | |||
| AF283301.1 | AAK73017.1 | |||
| AF283302.1 | AAM69373.1 | |||
| AF521133.1 | AAQ08823.1 | |||
| AK027897.1 | BAB55439.1 | |||
| AK301278.1 | ||||
| AK307663.1 | ||||
| AK315006.1 | ||||
| AL832317.1 | CAD38612.1 | |||
| BC000967.2 | AAH00967.2 | |||
| BC031258.1 | AAH31258.1 | |||
| BC043406.1 | AAH43406.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv7281n100 | Chr.8:109157344 - 109290856 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map