Assay Details
Target Gene Details
Entrez Gene ID: | 55284 |
Gene Name: | ubiquitin conjugating enzyme E2 W (putative) |
Gene Aliases: |
UBC-16, UBC16 |
Location: |
Chr.8:73780097-73878910 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UBE2W | NM_001001481.2 | 7 | 7181 | NP_001001481.2 |
| NM_001271015.1 | NP_001257944.1 | |||
| NM_018299.4 | 6 | 7148 | NP_060769.4 | |
| NR_073119.1 | 6 | 7169 | ||
| NR_073120.1 | 6 | 7129 | ||
| NR_073121.1 | 5 | 7045 | ||
| AK123531.1 | 1 | 1531 | ||
| BC010900.1 | AAH10900.1 |
More Information
Set Membership: |
|

Genomic Map