Assay Details
Target Gene Details
Entrez Gene ID: | 51115 |
Gene Name: | regulator of microtubule dynamics 1 |
Gene Aliases: |
CGI-90, FAM82B, RMD-1, RMD1 |
Location: |
Chr.8:86467116-86514348 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RMDN1 | XM_005250944.3 | XP_005251001.1 | ||
| XM_005250947.3 | XP_005251004.1 | |||
| XM_005250948.3 | XP_005251005.1 | |||
| XM_006716573.3 | XP_006716636.1 | |||
| XM_011517090.2 | XP_011515392.1 | |||
| XM_011517091.2 | XP_011515393.1 | |||
| XM_011517092.2 | XP_011515394.1 | |||
| XM_011517095.2 | XP_011515397.1 | |||
| XM_017013518.1 | XP_016869007.1 | |||
| XM_017013525.1 | XP_016869014.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1023510 | Chr.8:86416772 - 86862720 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs147549001] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map