Assay Details
Target Gene Details
Entrez Gene ID: | 79977 |
Gene Name: | grainyhead like transcription factor 2 |
Gene Aliases: |
BOM, DFNA28, ECTDS, TFCP2L3 |
Location: |
Chr.8:101492440-101678105 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GRHL2 | NM_024915.3 | NP_079191.2 | ||
| XM_011517305.2 | XP_011515607.1 | |||
| XM_011517306.2 | XP_011515608.1 | |||
| XM_011517307.2 | XP_011515609.1 | |||
| AK023844.1 | BAB14699.1 | |||
| AK292853.1 | ||||
| AK296817.1 | ||||
| AK310221.1 | ||||
| BC069618.1 | AAH69618.1 | |||
| BC069633.1 | AAH69633.1 | |||
| BC069638.1 | AAH69638.1 | |||
| BC129822.1 | ||||
| BC129823.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs78481884] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map