Assay Details
Target Gene Details
Entrez Gene ID: | 2339 |
Gene Name: | farnesyltransferase, CAAX box, alpha |
Gene Aliases: |
FPTA, PGGT1A, PTAR2 |
Location: |
Chr.8:43056299-43085788 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FNTA | NM_002027.2 | NP_002018.1 | ||
| NR_033698.1 | ||||
| AK123002.1 | ||||
| AK292121.1 | ||||
| AL698961.1 | ||||
| BC017029.2 | AAH17029.2 | |||
| BC084566.1 | AAH84566.1 | |||
| BI913631.1 | ||||
| BM686906.1 | ||||
| BT009854.1 | AAP88856.1 | |||
| L00634.1 | AAA35853.1 | |||
| L10413.1 | AAA86285.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2668009 | Chr.8:43065499 - 43066511 on Build GRCh38 | Deletion |
|
| esv3891387 | Chr.8:42998571 - 43918067 on Build GRCh38 | Gain |
|
| nsv831302 | Chr.8:43054340 - 43250377 on Build GRCh38 | Loss |
|
| nsv1032781 | Chr.8:42917251 - 43606341 on Build GRCh38 | Gain |
|
| esv2235385 | Chr.8:43065371 - 43066667 on Build GRCh38 | Deletion |
|
| esv2547171 | Chr.8:43064574 - 43066586 on Build GRCh38 | Deletion |
|
| esv3617025 | Chr.8:43065451 - 43066458 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map