Assay Details
Target Gene Details
Entrez Gene ID: | 157807 |
Gene Name: | clavesin 1 |
Gene Aliases: |
CRALBPL, RLBP1L1 |
Location: |
Chr.8:61057158-61501645 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLVS1 | XM_017013141.1 | XP_016868630.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv465697 | Chr.8:61051140 - 61113389 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map