Assay Details
Target Gene Details
Entrez Gene ID: | 10987 |
Gene Name: | COP9 signalosome subunit 5 |
Gene Aliases: |
CSN5, JAB1, MOV-34, SGN5 |
Location: |
Chr.8:67043079-67062327 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COPS5 | NM_006837.2 | NP_006828.2 | ||
| AB209134.1 | 6 | 4336 | BAD92371.1 | |
| AK056967.1 | ||||
| AK128519.1 | ||||
| BC001187.1 | AAH01187.1 | |||
| BC001859.2 | AAH01859.1 | |||
| BC007272.1 | AAH07272.1 | |||
| BX648542.1 | CAH10375.1 | |||
| CR541678.1 | CAG46479.1 | |||
| U65928.1 | AAB16847.1 | |||
| U70734.1 | AAD03468.1 |
More Information
Set Membership: |
|

Genomic Map