Assay Details
Target Gene Details
Entrez Gene ID: | 727677 |
Gene Name: | cancer susceptibility candidate 8 (non-protein coding) |
Gene Aliases: |
CARLo-1, LINC00860 |
Location: |
Chr.8:127289676-127482139 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CASC8 | NR_117100.1 | |||
| DQ515897.1 | ||||
| DQ515898.1 | ||||
| DQ515899.1 | ||||
| JX003864.1 | ||||
| JX003866.1 |
Target Gene Details
Entrez Gene ID: | 101805488 |
Gene Name: | colon cancer associated transcript 2 (non-protein coding) |
Gene Aliases: |
LINC00873, NCCP1 |
Location: |
Chr.8:127400399-127402150 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCAT2 | NR_109834.1 | 1 | 955 | |
| GQ911592.1 | 1 | 955 |
More Information
Set Membership: |
|

Genomic Map