Assay Details
Target Gene Details
Entrez Gene ID: | 56943 |
Gene Name: | ENY2, transcription and export complex 2 subunit |
Gene Aliases: |
DC6, Sus1, e(y)2 |
Location: |
Chr.8:109334323-109345960 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ENY2 | NM_001193557.1 | 5 | 2228 | NP_001180486.1 |
| NM_020189.5 | 5 | 2300 | NP_064574.1 | |
| NR_036471.1 | 5 | 2298 | ||
| NR_036472.1 | 5 | 2345 | ||
| AK095651.1 | 5 | 2344 | ||
| BU737295.1 | ||||
| BX537648.1 | 2 | 2124 |
More Information
Set Membership: |
|

Genomic Map