Assay Details
Target Gene Details
Entrez Gene ID: | 286135 |
Gene Name: | family with sequence similarity 183 member C, pseudogene |
Gene Aliases: |
- |
Location: |
Chr.8:29921513-29953609 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM183CP | NR_024473.1 | |||
| AL833525.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3616779 | Chr.8:29919111 - 29925745 on Build GRCh38 | Loss |
|

Genomic Map