Assay Details
Target Gene Details
Entrez Gene ID: | 7275 |
Gene Name: | tubby bipartite transcription factor |
Gene Aliases: |
RDOB, rd5 |
Location: |
Chr.11:8019180-8106107 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TUB | XM_005253109.3 | XP_005253166.1 | ||
| AK293528.1 |
More Information
Set Membership: |
|

Genomic Map