Assay Details
Target Gene Details
Entrez Gene ID: | 2348 |
Gene Name: | folate receptor 1 |
Gene Aliases: |
FBP, FOLR |
Location: |
Chr.11:72189558-72196323 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FOLR1 | NM_000802.3 | NP_000793.1 | ||
| NM_016724.2 | NP_057936.1 | |||
| NM_016725.2 | NP_057937.1 | |||
| AK223527.1 | BAD97247.1 | |||
| BC002947.2 | AAH02947.1 | |||
| J05013.1 | AAA35823.1 | |||
| M28099.1 | AAA35822.1 | |||
| X62753.1 | CAA44610.1 |
More Information
Set Membership: |
|

Genomic Map