Assay Details
Target Gene Details
Entrez Gene ID: | 100652768 |
Gene Name: | uncharacterized LOC100652768 |
Gene Aliases: |
- |
Location: |
Chr.11:117195613-117201914 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100652768 | NR_045215.1 | |||
| AK026219.1 |
Target Gene Details
Entrez Gene ID: | 6876 |
Gene Name: | transgelin |
Gene Aliases: |
SM22, SMCC, TAGLN1, WS3-10 |
Location: |
Chr.11:117199324-117204792 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TAGLN | NM_001001522.1 | NP_001001522.1 | ||
| NM_003186.3 | NP_003177.2 | |||
| AK223002.1 | BAD96722.1 | |||
| AK314043.1 | ||||
| BC004927.1 | AAH04927.1 | |||
| BC010946.1 | AAH10946.1 | |||
| BC093050.1 | AAH93050.1 | |||
| BI830888.1 | ||||
| CR749612.1 | 1 | 909 | CAH18406.1 | |
| D17409.1 | BAA21811.1 | |||
| M95787.1 | AAA58351.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv2186n54 | Chr.11:117188631 - 117257555 on Build GRCh38 | Loss |
|
| nsv524270 | Chr.11:117180449 - 117214554 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map