Assay Details
Target Gene Details
Entrez Gene ID: | 3784 |
Gene Name: | potassium voltage-gated channel subfamily Q member 1 |
Gene Aliases: |
ATFB1, ATFB3, JLNS1, KCNA8, KCNA9, KVLQT1, Kv1.9, Kv7.1, LQT, LQT1, RWS, SQT2, WRS |
Location: |
Chr.11:2444991-2849110 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNQ1 | NM_000218.2 | NP_000209.2 | ||
| NM_181798.1 | NP_861463.1 | |||
| AF000571.1 | AAC51776.1 | |||
| AF051426.1 | AAC05705.1 | |||
| AK129678.1 | ||||
| AK290618.1 | ||||
| AY114213.1 | AAM94040.1 | |||
| BC017074.1 | AAH17074.1 | |||
| BC113545.1 | ||||
| U89364.1 | AAC51781.1 |
Target Gene Details
Entrez Gene ID: | 10984 |
Gene Name: | KCNQ1 opposite strand/antisense transcript 1 (non-protein coding) |
Gene Aliases: |
KCNQ1-AS2, KCNQ10T1, Kncq1, KvDMR1, KvLQT1-AS, LIT1, NCRNA00012 |
Location: |
Chr.11:2608328-2699998 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNQ1OT1 | NR_002728.3 | 1 | 22658 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1037537 | Chr.11:2409367 - 2810311 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map