Assay Details
Target Gene Details
Entrez Gene ID: | 64231 |
Gene Name: | membrane spanning 4-domains A6A |
Gene Aliases: |
4SPAN3, 4SPAN3.2, CD20L3, CDA01, MS4A6, MST090, MSTP090 |
Location: |
Chr.11:60171607-60184666 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MS4A6A | NM_001247999.1 | 1 | 305 | NP_001234928.1 |
| NM_152852.2 | NP_690591.1 | |||
| XM_005274177.2 | 1 | 300 | XP_005274234.1 | |
| XM_006718658.2 | 1 | 128 | XP_006718721.1 | |
| XM_006718660.2 | 1 | 128 | XP_006718723.1 | |
| XM_006718661.2 | 1 | 128 | XP_006718724.1 | |
| XM_011545209.1 | 1 | 128 | XP_011543511.1 | |
| XM_017018125.1 | 1 | 128 | XP_016873614.1 | |
| AF172819.1 | AAQ13613.1 | |||
| AK291870.1 | ||||
| DC295707.1 | 1 | 306 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832174 | Chr.11:60141794 - 60306435 on Build GRCh38 | Gain |
|
| nsv555150 | Chr.11:60046814 - 60207927 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map