Assay Details
Target Gene Details
Entrez Gene ID: | 4607 |
Gene Name: | myosin binding protein C, cardiac |
Gene Aliases: |
CMD1MM, CMH4, FHC, LVNC10, MYBP-C |
Location: |
Chr.11:47331406-47352702 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MYBPC3 | NM_000256.3 | NP_000247.2 | ||
| AK291569.1 | ||||
| AK297843.1 | ||||
| AL691822.1 | ||||
| AU140784.1 | ||||
| BC136543.1 | ||||
| BC136546.1 | ||||
| BC142685.1 | ||||
| BC151211.1 | ||||
| EF560722.1 | ||||
| EU747718.1 | ||||
| EU747719.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832141 | Chr.11:47311629 - 47499441 on Build GRCh38 | Loss |
|
| nsv509404 | Chr.11:47335304 - 47371698 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map