Assay Details
Target Gene Details
Entrez Gene ID: | 101928069 |
Gene Name: | uncharacterized LOC101928069 |
Gene Aliases: |
- |
Location: |
Chr.11:66473490-66480249 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101928069 | NR_120586.1 | |||
| BQ223044.1 | ||||
| HY006924.1 | ||||
| HY022823.1 | 2 | 296 | ||
| HY130167.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522277 | Chr.11:66466552 - 66726354 on Build GRCh38 | Loss |
|
| nsv832193 | Chr.11:66352868 - 66511334 on Build GRCh38 | Loss |
|

Genomic Map