Assay Details
Target Gene Details
Entrez Gene ID: | 54860 |
Gene Name: | membrane spanning 4-domains A12 |
Gene Aliases: |
Ms4a10 |
Location: |
Chr.11:60492743-60507430 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MS4A12 | NM_001164470.1 | NP_001157942.1 | ||
| NM_017716.2 | NP_060186.2 | |||
| XM_011545117.2 | XP_011543419.1 | |||
| AK000224.1 | BAA91020.1 | |||
| BC029793.1 | AAH29793.1 | |||
| BP262076.1 | ||||
| BP262976.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv337 | Chr.11:60487120 - 60519417 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map