Assay Details
Target Gene Details
Entrez Gene ID: | 27122 |
Gene Name: | dickkopf WNT signaling pathway inhibitor 3 |
Gene Aliases: |
REIC, RIG |
Location: |
Chr.11:11962996-12009370 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DKK3 | NM_001018057.1 | NP_001018067.1 | ||
| NM_013253.4 | NP_037385.2 | |||
| NM_015881.5 | NP_056965.3 | |||
| XM_006718178.2 | XP_006718241.1 | |||
| XM_017017553.1 | XP_016873042.1 | |||
| XM_017017554.1 | XP_016873043.1 | |||
| XM_017017555.1 | XP_016873044.1 | |||
| AB033421.1 | BAA85488.1 | |||
| AB034203.1 | BAA90548.1 | |||
| AB057591.1 | BAB84360.1 | |||
| AF177396.1 | AAF02676.1 | |||
| AK092979.1 | ||||
| AK098756.1 | ||||
| AK225156.1 | ||||
| AK289897.1 | ||||
| AK295441.1 | ||||
| AK295539.1 | ||||
| AL535720.3 | ||||
| AY358378.1 | AAQ88744.1 | |||
| AY587550.1 | AAS86757.1 | |||
| BC007660.2 | AAH07660.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv483000 | Chr.11:10678454 - 12678453 on Build GRCh38 | Loss |
|
| nsv832067 | Chr.11:11777158 - 11976774 on Build GRCh38 | Gain |
|
| nsv553544 | Chr.11:11956026 - 11977482 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map