Assay Details
Target Gene Details
Entrez Gene ID: | 143879 |
Gene Name: | kelch repeat and BTB domain containing 3 |
Gene Aliases: |
BKLHD3 |
Location: |
Chr.11:106046801-106077765 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KBTBD3 | NM_152433.3 | NP_689646.2 | ||
| NM_198439.2 | NP_940841.1 | |||
| XM_006718767.3 | XP_006718830.1 | |||
| XM_011542618.2 | XP_011540920.1 | |||
| XM_011542619.2 | XP_011540921.1 | |||
| AK092993.1 | BAC04012.1 | |||
| AK289915.1 | ||||
| BC107691.1 | AAI07692.1 | |||
| BC117466.1 | ||||
| BX640672.1 | CAE45804.1 | |||
| BX647830.1 | ||||
| CD104687.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1052674 | Chr.11:104111970 - 107043401 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map