Assay Details
Target Gene Details
Entrez Gene ID: | 23365 |
Gene Name: | Rho guanine nucleotide exchange factor 12 |
Gene Aliases: |
LARG, PRO2792 |
Location: |
Chr.11:120336276-120489936 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARHGEF12 | NM_001198665.1 | NP_001185594.1 | ||
| NM_015313.2 | NP_056128.1 | |||
| XM_006718805.3 | XP_006718868.1 | |||
| XM_011542720.2 | XP_011541022.1 | |||
| XM_017017420.1 | XP_016872909.1 | |||
| XM_017017421.1 | XP_016872910.1 | |||
| AF180681.1 | AAF36817.1 | |||
| AK304316.1 | ||||
| BC063117.1 | AAH63117.1 | |||
| BX648515.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv529003 | Chr.11:120338677 - 120484521 on Build GRCh38 | Loss |
|
| nsv1050935 | Chr.11:120353835 - 120405664 on Build GRCh38 | Gain |
|
| dgv1287n100 | Chr.11:120359534 - 120428889 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map