Assay Details
Target Gene Details
Entrez Gene ID: | 105369438 |
Gene Name: | uncharacterized LOC105369438 |
Gene Aliases: |
- |
Location: |
Chr.11:94545332-94740355 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC105369438 | NR_135093.1 | |||
| NR_135094.1 | ||||
| NR_135096.1 | ||||
| BI522712.1 | ||||
| DA410022.1 |
Target Gene Details
Entrez Gene ID: | 143689 |
Gene Name: | piwi like RNA-mediated gene silencing 4 |
Gene Aliases: |
HIWI2, MIWI2 |
Location: |
Chr.11:94567308-94621421 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PIWIL4 | NM_152431.2 | NP_689644.2 | ||
| AB079366.1 | BAC81341.1 | |||
| AK093475.1 | BAC04179.1 | |||
| AK293616.1 | ||||
| BC031060.1 | AAH31060.1 | |||
| CR749642.1 | CAH18436.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2671948 | Chr.11:94349534 - 94645424 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map