Assay Details
Target Gene Details
Entrez Gene ID: | 219956 |
Gene Name: | olfactory receptor family 9 subfamily Q member 1 |
Gene Aliases: |
- |
Location: |
Chr.11:58023881-58181587 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR9Q1 | NM_001005212.3 | NP_001005212.1 | ||
| BC110819.1 | AAI10820.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832165 | Chr.11:58038552 - 58241604 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs116098240] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map