Assay Details
Target Gene Details
Entrez Gene ID: | 196074 |
Gene Name: | methyltransferase like 15 |
Gene Aliases: |
METT5D1 |
Location: |
Chr.11:28108251-28527041 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| METTL15 | NM_001113528.1 | NP_001107000.1 | ||
| NM_001297775.1 | NP_001284704.1 | |||
| NM_152636.2 | NP_689849.2 | |||
| XM_011519935.2 | XP_011518237.1 | |||
| XM_011519937.2 | XP_011518239.1 | |||
| XM_011519940.2 | XP_011518242.1 | |||
| XM_011519941.2 | XP_011518243.1 | |||
| XM_017017297.1 | XP_016872786.1 | |||
| XM_017017298.1 | XP_016872787.1 | |||
| XM_017017299.1 | 1 | 689 | XP_016872788.1 | |
| XM_017017300.1 | XP_016872789.1 | |||
| XM_017017301.1 | XP_016872790.1 | |||
| XM_017017303.1 | XP_016872792.1 | |||
| AK091298.1 | BAC03631.1 | |||
| AL832668.1 | ||||
| BC030997.1 | AAH30997.1 | |||
| BC105287.1 | AAI05288.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3891958 | Chr.11:28066977 - 28124180 on Build GRCh38 | Gain |
|
| esv3579455 | Chr.11:28097543 - 28156843 on Build GRCh38 | Loss |
|
| nsv832096 | Chr.11:27998888 - 28134039 on Build GRCh38 | Loss |
|
| nsv1040043 | Chr.11:28096904 - 28149137 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map