Assay Details
Target Gene Details
Entrez Gene ID: | 101054525 |
Gene Name: | progesterone receptor antisense RNA |
Gene Aliases: |
- |
Location: |
Chr.11:101129077-101159270 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101054525 | NR_073144.1 | |||
| FJ515872.1 | ||||
| FJ515873.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv556131 | Chr.11:101115046 - 101609286 on Build GRCh38 | Gain |
|

Genomic Map