Assay Details
Target Gene Details
Entrez Gene ID: | 114571 |
Gene Name: | solute carrier family 22 member 9 |
Gene Aliases: |
HOAT4, OAT4, OAT7, UST3H, ust3 |
Location: |
Chr.11:63368871-63410878 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC22A9 | NM_080866.2 | NP_543142.2 | ||
| XM_017017159.1 | XP_016872648.1 | |||
| XM_017017160.1 | XP_016872649.1 | |||
| AB062418.1 | BAB83517.1 | |||
| AB074812.1 | ||||
| AF440402.1 | AAP97316.1 | |||
| AJ295270.1 | CAC82910.1 | |||
| AK074246.1 | BAB85030.1 | |||
| BC022379.1 | AAH22379.1 | |||
| BC126288.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv555178 | Chr.11:63311270 - 63430458 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map