Assay Details
Target Gene Details
Entrez Gene ID: | 867 |
Gene Name: | Cbl proto-oncogene |
Gene Aliases: |
C-CBL, CBL2, FRA11B, NSLL, RNF55 |
Location: |
Chr.11:119206276-119308149 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CBL | NM_005188.3 | NP_005179.2 | ||
| XM_011543057.2 | XP_011541359.1 | |||
| XM_017018511.1 | XP_016874000.1 | |||
| BC132733.1 | ||||
| BC136463.1 | ||||
| BG722940.1 | ||||
| DB457271.2 | ||||
| X57110.1 | CAA40393.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3627889 | Chr.11:119243178 - 119245631 on Build GRCh38 | Loss |
|
| esv2665710 | Chr.11:119243113 - 119245775 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map