Assay Details
Target Gene Details
Entrez Gene ID: | 22800 |
Gene Name: | related RAS viral (r-ras) oncogene homolog 2 |
Gene Aliases: |
TC21 |
Location: |
Chr.11:14277920-14364506 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RRAS2 | NM_001102669.2 | NP_001096139.1 | ||
| NM_001177314.1 | NP_001170785.1 | |||
| NM_001177315.1 | NP_001170786.1 | |||
| NM_012250.5 | NP_036382.2 | |||
| XM_017017363.1 | XP_016872852.1 | |||
| AF493924.1 | AAM12638.1 | |||
| AK299606.1 | ||||
| AK300103.1 | ||||
| AK302033.1 | ||||
| AK309550.1 | ||||
| AK309683.1 | ||||
| AK313976.1 | ||||
| BC013106.1 | AAH13106.1 | |||
| BG428983.1 | ||||
| BP206021.1 | ||||
| BP206589.1 | ||||
| BX324335.2 | ||||
| DC368033.1 | ||||
| M31468.1 | AAA36545.1 |
More Information
Set Membership: |
|

Genomic Map