Assay Details
Target Gene Details
Entrez Gene ID: | 85458 |
Gene Name: | DIX domain containing 1 |
Gene Aliases: |
CCD1 |
Location: |
Chr.11:111927144-112022650 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DIXDC1 | NM_001037954.3 | NP_001033043.1 | ||
| NM_001278542.1 | NP_001265471.1 | |||
| XM_017018466.1 | XP_016873955.1 | |||
| XM_017018467.1 | XP_016873956.1 | |||
| BC035509.2 | AAH35509.1 | |||
| BC048294.1 | AAH48294.1 | |||
| BC064479.1 | AAH64479.1 | |||
| BC128600.1 | ||||
| DQ642016.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2745077 | Chr.11:111949712 - 112188677 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map