Assay Details
Target Gene Details
Entrez Gene ID: | 56946 |
Gene Name: | EMSY, BRCA2 interacting transcriptional repressor |
Gene Aliases: |
C11orf30, GL002 |
Location: |
Chr.11:76444928-76552901 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1050713 | Chr.11:76436041 - 76556073 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map