Assay Details
Target Gene Details
Entrez Gene ID: | 1374 |
Gene Name: | carnitine palmitoyltransferase 1A |
Gene Aliases: |
CPT1, CPT1-L, L-CPT1 |
Location: |
Chr.11:68754620-68844410 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CPT1A | NM_001031847.2 | NP_001027017.1 | ||
| NM_001876.3 | NP_001867.2 | |||
| XM_005273762.2 | XP_005273819.1 | |||
| XM_005273763.1 | XP_005273820.1 | |||
| XM_017017220.1 | XP_016872709.1 | |||
| AK025307.1 | ||||
| AK172798.1 | ||||
| AK309464.1 | ||||
| AK314301.1 | ||||
| BC000185.1 | AAH00185.1 | |||
| BT009791.1 | AAP88793.1 | |||
| L39211.1 | AAC41748.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3626741 | Chr.11:68778750 - 68779434 on Build GRCh38 | Loss |
|
| nsv509422 | Chr.11:68734013 - 68794964 on Build GRCh38 | Insertion |
|
| nsv825967 | Chr.11:68778904 - 68779630 on Build GRCh38 | Loss |
|
| nsv825968 | Chr.11:68779134 - 68779630 on Build GRCh38 | Loss |
|
| nsv428261 | Chr.11:68754616 - 68937835 on Build GRCh38 | Gain |
|
| esv2664873 | Chr.11:68778804 - 68779491 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map