Assay Details
Target Gene Details
Entrez Gene ID: | 3992 |
Gene Name: | fatty acid desaturase 1 |
Gene Aliases: |
D5D, FADS6, FADSD5, LLCDL1, TU12 |
Location: |
Chr.11:61799625-61817057 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FADS1 | NM_013402.4 | NP_037534.3 | ||
| XM_011545022.2 | XP_011543324.1 | |||
| AF084558.1 | AAG23120.1 | |||
| AF199596.1 | AAF29378.1 | |||
| AF226273.1 | AAF70457.1 | |||
| AK027427.1 | BAB55103.1 | |||
| AK027522.1 | BAB55173.1 | |||
| AK074754.1 | BAC11182.1 | |||
| AK074819.1 | BAC11229.1 | |||
| AK096275.1 | ||||
| AK222906.1 | BAD96626.1 | |||
| AK289552.1 | ||||
| AK298871.1 | ||||
| AK314199.1 | ||||
| AL512760.1 | CAC21679.1 | |||
| BC007846.2 | AAH07846.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv528697 | Chr.11:61690385 - 61919561 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map