Assay Details
Target Gene Details
Entrez Gene ID: | 196074 |
Gene Name: | methyltransferase like 15 |
Gene Aliases: |
METT5D1 |
Location: |
Chr.11:28108251-28527041 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| METTL15 |
Target Gene Details
Entrez Gene ID: | 102466876 |
Gene Name: | microRNA 8068 |
Gene Aliases: |
hsa-mir-8068 |
Location: |
Chr.11:28477481-28477548 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MIR8068 | NR_107035.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv1084n100 | Chr.11:28384121 - 28480307 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs78258445] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map