Assay Details
Target Gene Details
Entrez Gene ID: | 2542 |
Gene Name: | solute carrier family 37 member 4 |
Gene Aliases: |
G6PT1, G6PT2, G6PT3, GSD1b, GSD1c, GSD1d, PRO0685, TRG-19, TRG19 |
Location: |
Chr.11:119024351-119030906 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC37A4 | NM_001164277.1 | NP_001157749.1 | ||
| NM_001164278.1 | NP_001157750.1 | |||
| NM_001164279.1 | NP_001157751.1 | |||
| NM_001164280.1 | NP_001157752.1 | |||
| NM_001467.5 | NP_001458.1 | |||
| AB209026.1 | 1 | 2784 | BAD92263.1 | |
| AF110819.1 | AAF37735.1 | |||
| AF110820.1 | AAF37736.1 | |||
| AF110821.1 | 2 | 495 | ||
| AF110822.1 | ||||
| AF111852.1 | 1 | 928 | AAF16691.1 | |
| AK289642.1 | ||||
| AK294993.1 | ||||
| AK298395.1 | ||||
| AK300646.1 | ||||
| AK312628.1 | ||||
| AY423732.1 | AAS00495.1 | |||
| BC002400.2 | AAH02400.1 | |||
| BC003589.1 | AAH03589.1 | |||
| BC014663.1 | AAH14663.1 | |||
| BC015650.2 | AAH15650.1 | |||
| BC064563.1 | AAH64563.1 | |||
| CR456733.1 | CAG33014.1 | |||
| Y15409.1 | CAA75608.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv952744 | Chr.11:119024091 - 119030790 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map