Assay Details
Target Gene Details
Entrez Gene ID: | 154810 |
Gene Name: | angiomotin like 1 |
Gene Aliases: |
JEAP |
Location: |
Chr.11:94706418-94876753 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AMOTL1 | XM_005273798.4 | XP_005273855.1 | ||
| XM_006718772.3 | XP_006718835.1 | |||
| XM_011542626.2 | XP_011540928.1 |
Target Gene Details
Entrez Gene ID: | 105369438 |
Gene Name: | uncharacterized LOC105369438 |
Gene Aliases: |
- |
Location: |
Chr.11:94545332-94740355 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC105369438 | NR_135093.1 | |||
| NR_135094.1 | ||||
| NR_135096.1 | ||||
| BI522712.1 | ||||
| DA410022.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3627327 | Chr.11:94717955 - 94726723 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map