Assay Details
Target Gene Details
Entrez Gene ID: | 5866 |
Gene Name: | RAB3A interacting protein like 1 |
Gene Aliases: |
GRAB |
Location: |
Chr.11:61897296-61946312 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAB3IL1 | NM_001271686.1 | NP_001258615.1 | ||
| NM_013401.3 | NP_037533.2 | |||
| XM_005274140.4 | XP_005274197.1 | |||
| XM_006718647.3 | XP_006718710.1 | |||
| XM_011545196.2 | XP_011543498.1 | |||
| XM_011545197.2 | XP_011543499.1 | |||
| XM_011545198.2 | XP_011543500.1 | |||
| XM_011545199.2 | XP_011543501.1 | |||
| XM_011545201.2 | XP_011543503.1 | |||
| BC022239.1 | AAH22239.1 | |||
| BC051820.1 | AAH51820.1 | |||
| BQ946081.1 | ||||
| DR004757.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv528697 | Chr.11:61690385 - 61919561 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map